siRNA Details
| Gene Symbol: ZNF19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006961.3 | NP_008892.2 | 6 | 1212 | ||
| GenBank mRNA | AK055567.1 | - | 5 | 1093 | ||
| AK057330.1 | - | 5 | 1387 | |||
| AK292260.1 | - | 6 | 1410 | |||
| AL834237.1 | - | 1 | 586 | |||
| BC047091.1 | - | 6 | 1199 | 5270853 | ||
| X52343.1 | - | 1 | 127 | |||

ZNF19
Genomic Map