siRNA Details
| Gene Symbol: ZNF19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006961.3 | NP_008892.2 | 6 | 1420 | ||
| GenBank mRNA | AK055567.1 | - | 5 | 1301 | ||
| AK057330.1 | - | 5 | 1595 | |||
| AK292260.1 | - | 6 | 1618 | |||
| AL834237.1 | - | 1 | 794 | |||
| BC047091.1 | - | 6 | 1407 | 5270853 | ||

ZNF19
Genomic Map