siRNA Details
| Gene Symbol: ZNF19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006961.3 | NP_008892.2 | 6 | 2514 | ||
| GenBank mRNA | AL834237.1 | - | 1 | 1888 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF19| Gene Symbol: ZNF19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006961.3 | NP_008892.2 | 6 | 2514 | ||
| GenBank mRNA | AL834237.1 | - | 1 | 1888 | ||