siRNA Details
| Gene Symbol: ZNF24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006965.3 | NP_008896.2 | 4 | 2308 | ||
| XM_005258341.3 | XP_005258398.1 | 4 | 2295 | |||
| GenBank mRNA | U68536.1 | - | 4 | 2278 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF24| Gene Symbol: ZNF24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006965.3 | NP_008896.2 | 4 | 2308 | ||
| XM_005258341.3 | XP_005258398.1 | 4 | 2295 | |||
| GenBank mRNA | U68536.1 | - | 4 | 2278 | ||