siRNA Details
| Gene Symbol: ZNF197 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005264783.4 | XP_005264840.1 | 5 | 885 | ||
| XM_017005495.1 | XP_016860984.1 | 5 | 857 | |||
| GenBank mRNA | AF011573.1 | - | 4 | 672 | ||
| AK314196.1 | - | 5 | 853 | |||
| AY074878.1 | - | 5 | 853 | |||
| AY261677.1 | - | 4 | 686 | |||
| AY279353.1 | - | 5 | 874 | |||
| BC017584.1 | - | 4 | 838 | |||
| Z21707.1 | - | 4 | 584 | |||

ZNF197
Genomic Map