siRNA Details
| Gene Symbol: SSR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308197.1 | NP_001295126.1 | 5 | 910 | ||
| NM_001308204.1 | NP_001295133.1 | 5 | 736 | |||
| NM_001308205.1 | NP_001295134.1 | 5 | 760 | |||
| NM_007107.4 | NP_009038.1 | 5 | 871 | |||
| GenBank mRNA | AF110647.1 | - | 5 | 816 | ||
| AK304364.1 | - | 5 | 808 | |||
| BC017203.1 | - | 5 | 817 | 3618589 | ||

SSR3
Genomic Map