siRNA Details
| Gene Symbol: FKBP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001284341.1 | NP_001271270.1 | 6 | 1132 | ||
| NM_001284343.1 | NP_001271272.1 | 2 | 210 | |||
| NM_007270.4 | NP_009201.2 | 5 | 973 | |||
| XM_011515115.2 | XP_011513417.1 | 5 | 973 | |||
| GenBank mRNA | AF089745.1 | - | 5 | 644 | ||
| AK075331.1 | - | 5 | 937 | |||
| AK128597.1 | - | 6 | 914 | |||
| AK293440.1 | - | 2 | 188 | |||
| AK293954.1 | - | 4 | 546 | |||
| AK294266.1 | - | 4 | 757 | |||
| AK300328.1 | - | 6 | 1095 | |||
| BC101723.1 | - | 5 | 901 | 8069229 | ||
| BC112053.1 | - | 5 | 901 | 8327521 | ||
| BQ688269.1 | - | 2 | 210 | |||

FKBP9
Genomic Map