siRNA Details
| Gene Symbol: SLC35A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271684.1 | NP_001258613.1 | 6 | 1860 | ||
| NM_001271685.1 | NP_001258614.1 | 8 | 1948 | |||
| NM_012243.2 | NP_036375.1 | 8 | 2113 | |||
| XM_005270691.4 | XP_005270748.1 | 9 | 2634 | |||
| XM_011541135.2 | XP_011539437.1 | 7 | 1967 | |||
| XM_011541136.2 | XP_011539438.1 | 9 | 2030 | |||
| XM_011541137.2 | XP_011539439.1 | 7 | 1956 | |||
| XM_011541138.2 | XP_011539440.1 | 6 | 1846 | |||
| XM_017000869.1 | XP_016856358.1 | 7 | 2412 | |||
| XM_017000870.1 | XP_016856359.1 | 7 | 2401 | |||
| XM_017000871.1 | XP_016856360.1 | 6 | 2278 | |||
| XM_017000872.1 | XP_016856361.1 | 7 | 2380 | |||
| GenBank mRNA | AB021981.1 | - | 8 | 1918 | ||
| AK024989.1 | - | 7 | 1679 | |||
| BC005136.2 | - | 6 | 1643 | 3505772 | ||
| CR749816.1 | - | 8 | 1948 | |||

SLC35A3
Genomic Map