siRNA Details
| Gene Symbol: CLEC5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301167.1 | NP_001288096.1 | 6 | 947 | ||
| NM_013252.2 | NP_037384.1 | 7 | 992 | |||
| XM_011515995.2 | XP_011514297.1 | 5 | 879 | |||
| XM_017011915.1 | XP_016867404.1 | 6 | 950 | |||
| XM_017011917.1 | XP_016867406.1 | 6 | 945 | |||
| GenBank mRNA | AJ271684.1 | - | 7 | 992 | ||
| AK295200.1 | - | 6 | 884 | |||
| AK307095.1 | - | 5 | 810 | |||

CLEC5A
Genomic Map