siRNA Details
| Gene Symbol: SLC30A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013309.5 | NP_037441.2 | 7 | 1422 | ||
| XM_011521997.2 | XP_011520299.1 | 7 | 1926 | |||
| XM_017022560.1 | XP_016878049.1 | 6 | 1228 | |||
| GenBank mRNA | AF025409.1 | - | 6 | 1110 | ||
| AK290874.1 | - | 6 | 1228 | |||
| AY029482.1 | - | 3 | 387 | |||
| BC026089.2 | - | 7 | 1422 | 4829839 | ||

SLC30A4
Genomic Map