siRNA Details
| Gene Symbol: NUAK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030952.2 | NP_112214.2 | 2 | 467 | ||
| GenBank mRNA | AK074830.1 | - | 2 | 421 | ||
| AK303577.1 | - | 2 | 467 | |||
| AL136891.1 | - | 2 | 431 | |||
| BC017306.2 | - | 2 | 403 | 5088037 | ||

NUAK2
Genomic Map