siRNA Details
| Gene Symbol: PYCR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271681.1 | NP_001258610.1 | 6 | 1113 | ||
| NM_013328.3 | NP_037460.2 | 7 | 1335 | |||
| GenBank mRNA | AK001508.1 | - | 5 | 2714 | ||
| AK291913.1 | - | 7 | 1170 | |||
| AL834217.1 | - | 3 | 1068 | |||
| BC014868.1 | - | 7 | 1226 | 3919161 | ||
| BC020553.1 | - | 7 | 1260 | 2901360 | ||

PYCR2
Genomic Map