siRNA Details
| Gene Symbol: ZNF221 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001297588.1 | NP_001284517.1 | 5 | 610 | ||
| NM_001297589.1 | NP_001284518.1 | 5 | 620 | |||
| NM_013359.2 | NP_037491.2 | 6 | 758 | |||
| XM_017027232.1 | XP_016882721.1 | 5 | 637 | |||
| XM_017027233.1 | XP_016882722.1 | 4 | 554 | |||
| GenBank mRNA | AF187987.1 | - | 6 | 758 | ||
| AK314207.1 | - | 5 | 605 | |||
| BC111981.1 | - | 5 | 513 | 8327449 | ||
| BC113426.1 | - | 5 | 513 | 8322478 | ||
| BC143659.1 | - | 6 | 661 | |||

ZNF221
Genomic Map