siRNA Details
| Gene Symbol: ZNF223 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013361.5 | NP_037493.3 | 5 | 2174 | ||
| XM_017027258.1 | XP_016882747.1 | 5 | 2419 | |||
| XM_017027259.1 | XP_016882748.1 | 5 | 2474 | |||
| XM_017027260.1 | XP_016882749.1 | 2 | 3393 | |||
| GenBank mRNA | BC022466.1 | - | 5 | 2174 | 4793666 | |

ZNF223
Genomic Map