siRNA Details
| Gene Symbol: ZNF225 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321685.1 | NP_001308614.1 | 6 | 1605 | ||
| NM_013362.3 | NP_037494.2 | 5 | 1440 | |||
| XM_005259223.3 | XP_005259280.1 | 2 | 986 | |||
| XM_011527285.2 | XP_011525587.1 | 5 | 1460 | |||
| XM_011527286.2 | XP_011525588.1 | 5 | 2104 | |||
| GenBank mRNA | AF187991.1 | - | 5 | 1414 | ||
| AK223477.1 | - | 5 | 1359 | |||
| AK292437.1 | - | 5 | 1444 | |||
| AK302106.1 | - | 4 | 1156 | |||
| AK313301.1 | - | 4 | 1447 | |||
| BC108912.2 | - | 4 | 1237 | 40027157 | ||

ZNF225
Genomic Map