siRNA Details
| Gene Symbol: SLC25A24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013386.4 | NP_037518.3 | 5, 6 | 897 | ||
| NM_213651.2 | NP_998816.1 | 5, 6 | 808 | |||
| XM_017001104.1 | XP_016856593.1 | 1, 2 | 153 | |||
| GenBank mRNA | AF123303.1 | - | 5, 6 | 656 | ||
| AJ619961.1 | - | 5, 6 | 740 | |||
| AJ619987.1 | - | 5, 6 | 686 | |||
| AK292567.1 | - | 5, 6 | 808 | |||
| AK304165.1 | - | 5, 6 | 891 | |||
| AK316304.1 | - | 5, 6 | 851 | |||
| AK316506.1 | - | 5, 6 | 719 | |||
| BC014519.1 | - | 5, 6 | 753 | 3842039 | ||
| BC068561.1 | - | 5, 6 | 786 | 5259336 | ||
| BQ685792.1 | - | 5, 6 | 814 | |||

SLC25A24
Genomic Map