siRNA Details
| Gene Symbol: VCL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003373.3 | NP_003364.1 | 3, 4 | 486 | ||
| NM_014000.2 | NP_054706.1 | 3, 4 | 486 | |||
| GenBank mRNA | AK126978.1 | - | 3, 4 | 741 | ||
| BC039174.1 | - | 3, 4 | 473 | 4520338 | ||
| BX537994.1 | - | 5, 6 | 351 | |||
| EU794673.1 | - | 3, 4 | 486 | |||
| M33308.1 | - | 3, 4 | 442 | |||

VCL
Genomic Map