siRNA Details
| Gene Symbol: SLC27A6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017372.2 | NP_001017372.1 | 5 | 2012 | ||
| NM_001317984.1 | NP_001304913.1 | 6 | 1836 | |||
| NM_014031.4 | NP_054750.1 | 6 | 1787 | |||
| GenBank mRNA | AF064254.1 | - | 5 | 1154 | ||
| AK313456.1 | - | 6 | 1435 | |||
| AL833117.1 | - | 5 | 1585 | |||
| BC041945.1 | - | 5 | 2011 | 5301306 | ||
| BX537383.1 | - | 6 | 1397 | |||
| CR457129.1 | - | 5 | 1001 | |||

SLC27A6
Genomic Map