siRNA Details
| Gene Symbol: NDUFA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318195.1 | NP_001305124.1 | 2 | 328 | ||
| NM_014222.2 | NP_055037.1 | 2 | 328 | |||
| GenBank mRNA | AF044953.1 | - | 2 | 253 | ||
| AK301520.1 | - | 2 | 296 | |||
| AK314135.1 | - | 2 | 288 | |||
| BC001016.2 | - | 2 | 232 | 3345138 | ||
| BU602971.1 | - | 2 | 330 | |||

NDUFA8
Genomic Map