siRNA Details
| Gene Symbol: SLC5A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014227.2 | NP_055042.1 | 14 | 1714 | ||
| XM_006724308.3 | XP_006724371.1 | 15 | 1965 | |||
| XM_011530342.2 | XP_011528644.1 | 16 | 2083 | |||
| XM_011530343.2 | XP_011528645.1 | 14 | 1629 | |||
| XM_011530344.2 | XP_011528646.1 | 13 | 1594 | |||
| XM_017028920.1 | XP_016884409.1 | 16 | 2044 | |||
| GenBank mRNA | AJ133127.1 | - | 14 | 1728 | ||
| GQ292716.1 | - | 3 | 442 | |||
| JQ340171.1 | - | 3 | 442 | |||
| JQ519903.1 | - | 3 | 442 | |||

SLC5A4
Genomic Map