siRNA Details
| Gene Symbol: IL17RA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289905.1 | NP_001276834.1 | 12 | 2991 | ||
| NM_014339.6 | NP_055154.3 | 13 | 3093 | |||
| GenBank mRNA | BC011624.2 | - | 13 | 3037 | 4053746 | |
| U58917.1 | - | 13 | 2992 | |||

IL17RA
Genomic Map