siRNA Details
| Gene Symbol: HIBCH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014362.3 | NP_055177.2 | 14 | 1465 | ||
| NM_198047.2 | NP_932164.1 | 13 | 1431 | |||
| XM_011510954.1 | XP_011509256.1 | 9 | 865 | |||
| GenBank mRNA | AK222979.1 | - | 14 | 1189 | ||
| AK223023.1 | - | 14 | 1189 | |||
| AK308042.1 | - | 14 | 1189 | |||
| BC005190.1 | - | 13 | 1208 | |||
| U66669.1 | - | 14 | 1218 | |||

HIBCH
Genomic Map