siRNA Details
| Gene Symbol: SND1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014390.2 | NP_055205.2 | 8 | 1104 | ||
| XM_017011987.1 | XP_016867476.1 | 8 | 1133 | |||
| GenBank mRNA | AB209510.1 | - | 8 | 1042 | ||
| AK096583.1 | - | 9 | 1029 | |||
| AK304181.1 | - | 8 | 996 | |||
| AK312313.1 | - | 8 | 1058 | |||
| BC017180.2 | - | 8 | 1060 | 3345037 | ||
| BT009785.1 | - | 8 | 878 | |||
| U22055.1 | - | 8 | 1070 | |||

SND1
Genomic Map