siRNA Details
| Gene Symbol: GYPE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002102.3 | NP_002093.2 | 3 | 247 | ||
| NM_198682.2 | NP_941391.2 | 3 | 247 | |||
| XM_017008139.1 | XP_016863628.1 | 4 | 4174 | |||
| XM_017008140.1 | XP_016863629.1 | 4 | 4174 | |||
| GenBank mRNA | BC017864.1 | - | 3 | 243 | 4686481 | |
| BC064396.1 | - | 3 | 204 | |||
| BG528203.1 | - | 3 | 243 | |||
| BX091225.1 | - | 3 | 215 | |||
| M29610.1 | - | 3 | 247 | |||

GYPE
Genomic Map