siRNA Details
| Gene Symbol: ZNF10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015394.4 | NP_056209.2 | 5 | 3402 | ||
| GenBank mRNA | BC024182.2 | - | 5 | 3403 | 4823159 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF10| Gene Symbol: ZNF10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015394.4 | NP_056209.2 | 5 | 3402 | ||
| GenBank mRNA | BC024182.2 | - | 5 | 3403 | 4823159 | |