siRNA Details
| Gene Symbol: CD207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015717.4 | NP_056532.4 | 6 | 1124 | ||
| XM_011532874.1 | XP_011531176.1 | 7 | 1312 | |||
| GenBank mRNA | AJ242859.1 | - | 6 | 1124 | ||
| BC022278.1 | - | 6 | 1117 | 4692155 | ||

CD207
Genomic Map