siRNA Details
| Gene Symbol: TAF9B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015975.4 | NP_057059.2 | 7 | 2595 | ||
| GenBank mRNA | BC009566.1 | - | 7 | 2548 | 3996687 | |
| BC071649.1 | - | 7 | 2532 | 5246715 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TAF9B| Gene Symbol: TAF9B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015975.4 | NP_057059.2 | 7 | 2595 | ||
| GenBank mRNA | BC009566.1 | - | 7 | 2548 | 3996687 | |
| BC071649.1 | - | 7 | 2532 | 5246715 | ||