siRNA Details
| Gene Symbol: GLRX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243399.1 | NP_001230328.1 | 4 | 726 | ||
| NM_001319291.1 | NP_001306220.1 | 4 | 307 | |||
| NM_016066.4 | NP_057150.2 | 4 | 884 | |||
| NM_197962.2 | NP_932066.1 | 4 | 501 | |||
| XM_017001397.1 | XP_016856886.1 | 4 | 434 | |||
| GenBank mRNA | AF132495.2 | - | 4 | 416 | ||
| AF151891.1 | - | 4 | 391 | |||
| AF290514.1 | - | 4 | 884 | |||
| AY038988.1 | - | 4 | 423 | |||
| BC009669.1 | - | 4 | 416 | |||
| BC028113.1 | - | 4 | 407 | 5395272 | ||
| BG182176.1 | - | 3 | 293 | |||
| BM887926.1 | - | 4 | 307 | |||
| BU157087.1 | - | 456 | ||||

GLRX2
Genomic Map