siRNA Details
| Gene Symbol: PDGFC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016205.2 | NP_057289.1 | 6 | 2062 | ||
| NR_036641.1 | 7 | 2123 | ||||
| XM_011532124.2 | XP_011530426.1 | 8 | 1742 | |||
| XM_017008455.1 | XP_016863944.1 | 6 | 1691 | |||
| XM_017008456.1 | XP_016863945.1 | 6 | 1835 | |||
| GenBank mRNA | AF091434.1 | - | 6 | 2062 | ||
| AF244813.1 | - | 6 | 1759 | |||
| AF260738.1 | - | 6 | 1734 | |||
| AY358493.1 | - | 6 | 1855 | |||
| BC094746.1 | - | 6 | 1662 | |||

PDGFC
Genomic Map