siRNA Details
| Gene Symbol: ZNF44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164276.1 | NP_001157748.1 | 5 | 802 | ||
| NM_016264.3 | NP_057348.3 | 4 | 658 | |||
| XM_005259934.3 | XP_005259991.1 | 4 | 668 | |||
| XM_011528062.2 | XP_011526364.1 | 5 | 767 | |||
| XM_011528064.1 | XP_011526366.1 | 4 | 1083 | |||
| XM_011528066.1 | XP_011526368.1 | 3 | 956 | |||
| XM_017026870.1 | XP_016882359.1 | 4 | 762 | |||
| XM_017026871.1 | XP_016882360.1 | 5 | 801 | |||
| GenBank mRNA | AB021642.1 | - | 4 | 593 | ||
| AK297528.1 | - | 5 | 716 | |||
| BC136502.1 | - | 5 | 697 | 9020491 | ||
| BE766488.1 | - | 1 | 152 | |||
| CN364680.1 | - | 4 | 637 | |||
| DA815414.1 | - | 4 | 561 | |||

ZNF44
Genomic Map