siRNA Details
| Gene Symbol: RNF138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191324.1 | NP_001178253.2 | 7 | 1407 | ||
| NM_016271.4 | NP_057355.2 | 8 | 1694 | |||
| NM_198128.2 | NP_937761.1 | 5 | 1125 | |||
| XM_005258285.1 | XP_005258342.1 | 7 | 1471 | |||
| XM_005258286.1 | XP_005258343.1 | 6 | 1412 | |||
| GenBank mRNA | AF162680.3 | - | 8 | 1454 | ||
| AK023579.1 | - | 5 | 1043 | |||
| AL133557.1 | - | 6 | 1047 | |||
| BC018107.1 | - | 8 | 1335 | 3914953 | ||

RNF138
Genomic Map