siRNA Details
Gene Symbol: RNF138 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191324.1 | NP_001178253.2 | 7 | 2202 | ||
NM_016271.4 | NP_057355.2 | 8 | 2489 | |||
NM_198128.2 | NP_937761.1 | 5 | 1920 | |||
XM_005258285.1 | XP_005258342.1 | 7 | 2266 | |||
XM_005258286.1 | XP_005258343.1 | 6 | 2207 | |||
GenBank mRNA | AF162680.3 | - | 8 | 2249 | ||
AK023579.1 | - | 5 | 1838 | |||
AL133557.1 | - | 6 | 1842 | |||
BC018107.1 | - | 8 | 2130 | 3914953 |
Gene Symbol: RNF138P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001575.1 | 1 | 5665 | |||
GenBank mRNA | BC040295.1 | - | 1 | 5669 |
