siRNA Details
| Gene Symbol: UIMC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199297.2 | NP_001186226.1 | 7 | 1321 | ||
| NM_001199298.1 | NP_001186227.1 | 6 | 1096 | |||
| NM_016290.4 | NP_057374.3 | 6 | 1110 | |||
| XM_005265930.2 | XP_005265987.1 | 6 | 1084 | |||
| XM_006714871.2 | XP_006714934.1 | 6 | 1059 | |||
| XM_011534568.2 | XP_011532870.1 | 6 | 1075 | |||
| XM_011534570.1 | XP_011532872.1 | 6 | 1075 | |||
| XM_017009574.1 | XP_016865063.1 | 5 | 975 | |||
| GenBank mRNA | AF349313.1 | - | 6 | 1052 | ||
| AK023044.1 | - | 7 | 1098 | |||
| BC006078.1 | - | 6 | 1017 | 3659756 | ||
| BX537376.1 | - | 6 | 1110 | |||

UIMC1
Genomic Map