siRNA Details
| Gene Symbol: NUP98 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005387.6 | NP_005378.4 | 14 | 2055 | ||
| NM_016320.4 | NP_057404.2 | 14 | 2004 | |||
| NM_139131.4 | NP_624357.1 | 14 | 2004 | |||
| NM_139132.3 | NP_624358.2 | 14 | 2004 | |||
| GenBank mRNA | AB040538.1 | - | 14 | 1810 | ||
| AF071076.1 | - | 14 | 1707 | |||
| AF071077.1 | - | 14 | 1707 | |||
| AF231130.1 | - | 14 | 1727 | |||
| AK292882.1 | - | 14 | 1795 | |||
| AK301512.1 | - | 14 | 1463 | |||
| BC041136.1 | - | 14 | 1753 | 5179730 | ||
| U41815.1 | - | 14 | 1727 | |||

NUP98
Genomic Map