siRNA Details
| Gene Symbol: TRAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317747.1 | NP_001304676.1 | 5 | 1631 | ||
| NM_016388.3 | NP_057472.2 | 6 | 1742 | |||
| GenBank mRNA | AF161547.1 | - | 6 | 1656 | ||
| AJ224878.1 | - | 6 | 1641 | |||
| AJ240085.1 | - | 5 | 1557 | |||
| AV735609.1 | - | 1 | 308 | |||
| BC025713.1 | - | 6 | 1648 | 5227396 | ||

TRAT1
Genomic Map