siRNA Details
| Gene Symbol: IL1RAPL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017416.1 | NP_059112.1 | 3 | 1055 | ||
| XM_017029403.1 | XP_016884892.1 | 3 | 473 | |||
| GenBank mRNA | AF181285.1 | - | 1 | 217 | ||
| AF212016.1 | - | 2 | 299 | |||
| AF284436.1 | - | 3 | 450 | |||
| AJ272208.1 | - | 3 | 1055 | |||
| AJ290436.1 | - | 2 | 318 | |||
| AK290196.1 | - | 3 | 1171 | |||
| BC104784.1 | - | 3 | 347 | 8143787 | ||
| BC104786.1 | - | 3 | 347 | 8143789 | ||

IL1RAPL2
Genomic Map