siRNA Details
| Gene Symbol: PNRC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017761.3 | NP_060231.1 | 3 | 2016 | ||
| XM_017001691.1 | XP_016857180.1 | 3 | 1921 | |||
| GenBank mRNA | AF374386.1 | - | 3 | 1935 | ||
| AK000319.1 | - | 2 | 1737 | |||
| AK024774.1 | - | 1 | 1436 | |||
| BC001959.1 | - | 3 | 1983 | 3536977 | ||
| BC078177.1 | - | 3 | 1963 | 6526331 | ||
| BC085018.1 | - | 3 | 1986 | 6650445 | ||

PNRC2
Genomic Map