siRNA Details
| Gene Symbol: ZNF416 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017879.1 | NP_060349.1 | 4 | 2224 | ||
| XM_005259051.3 | XP_005259108.1 | 3 | 2160 | |||
| GenBank mRNA | BC000130.1 | - | 4 | 2224 | 2901256 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF416| Gene Symbol: ZNF416 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017879.1 | NP_060349.1 | 4 | 2224 | ||
| XM_005259051.3 | XP_005259108.1 | 3 | 2160 | |||
| GenBank mRNA | BC000130.1 | - | 4 | 2224 | 2901256 | |