siRNA Details
Gene Symbol: IFT57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018010.3 | NP_060480.1 | 11 | 2218 | ||
GenBank mRNA | AF139576.1 | - | 11 | 2048 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: IFT57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018010.3 | NP_060480.1 | 11 | 2218 | ||
GenBank mRNA | AF139576.1 | - | 11 | 2048 |