siRNA Details
| Gene Symbol: KDM4D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018039.2 | NP_060509.2 | 3 | 2507 | ||
| GenBank mRNA | AK001113.1 | - | 1 | 1278 | ||
| AK056162.1 | - | 1 | 1919 | |||
| AL137545.1 | - | 1 | 495 | |||
| BC119010.1 | - | 1 | 1876 | 40085404 | ||
| BC122858.1 | - | 1 | 1876 | 40085403 | ||

KDM4D
Genomic Map