siRNA Details
| Gene Symbol: TRIM68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304496.1 | NP_001291425.1 | 6 | 1759 | ||
| NM_018073.7 | NP_060543.5 | 7 | 2242 | |||
| GenBank mRNA | AK001231.1 | - | 4 | 1447 | ||
| AK022923.1 | - | 4 | 1412 | |||
| AK056523.1 | - | 7 | 2204 | |||
| AL832730.1 | - | 7 | 2192 | |||

TRIM68
Genomic Map