siRNA Details
| Gene Symbol: FOXJ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018416.2 | NP_060886.1 | 11 | 3288 | ||
| XM_011520760.2 | XP_011519062.1 | 12 | 3462 | |||
| XM_011520761.2 | XP_011519063.1 | 13 | 3563 | |||
| XM_017019640.1 | XP_016875129.1 | 11 | 2126 | |||
| GenBank mRNA | AF155132.1 | - | 11 | 2692 | ||
| AK092529.1 | - | 1 | 229 | |||
| AL161978.1 | - | 3 | 832 | |||
| BC126396.1 | - | 11 | 2258 | 8992112 | ||
| BC136305.1 | - | 11 | 2258 | 9020292 | ||

FOXJ2
Genomic Map