siRNA Details
| Gene Symbol: SLC22A11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001307985.1 | NP_001294914.1 | 8 | 2033 | ||
| NM_018484.3 | NP_060954.1 | 10 | 2357 | |||
| XM_011545167.1 | XP_011543469.1 | 9 | 2007 | |||
| GenBank mRNA | AB026116.1 | - | 10 | 2010 | ||
| AK075224.1 | - | 10 | 1703 | |||
| AK129930.1 | - | 8 | 1945 | |||
| AK290791.1 | - | 10 | 2340 | |||
| BC034384.1 | - | 10 | 2332 | 5190509 | ||

SLC22A11
Genomic Map