siRNA Details
| Gene Symbol: TREM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242590.2 | NP_001229519.1 | 3 | 694 | ||
| NM_018643.4 | NP_061113.1 | 4 | 887 | |||
| NR_136332.1 | 4 | 887 | ||||
| XM_017010956.1 | XP_016866445.1 | 4 | 827 | |||
| XM_017010957.1 | XP_016866446.1 | 3 | 631 | |||
| GenBank mRNA | AF196329.1 | - | 4 | 844 | ||
| AF287008.1 | - | 4 | 861 | |||
| AK292162.1 | - | 4 | 824 | |||
| AK301519.1 | - | 3 | 631 | |||
| BC017773.1 | - | 4 | 842 | 4692680 | ||
| DA362458.1 | - | 3 | 334 | |||

TREM1
Genomic Map