siRNA Details
| Gene Symbol: ZNF26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256279.1 | NP_001243208.1 | 5 | 3045 | ||
| NM_001256280.1 | NP_001243209.1 | 3 | 2850 | |||
| NM_019591.3 | NP_062537.2 | 4 | 2946 | |||
| XM_005266182.3 | XP_005266239.1 | 5 | 3033 | |||
| XM_005266183.3 | XP_005266240.1 | 6 | 3236 | |||
| XM_011534829.1 | XP_011533131.1 | 5 | 3149 | |||
| XM_017019920.1 | XP_016875409.1 | 6 | 3669 | |||
| XM_017019921.1 | XP_016875410.1 | 6 | 3132 | |||
| XM_017019922.1 | XP_016875411.1 | 5 | 4018 | |||
| XM_017019923.1 | XP_016875412.1 | 5 | 3949 | |||
| GenBank mRNA | AK000762.1 | - | 1 | 2149 | ||
| AL833057.1 | - | 1 | 1723 | |||
| BC008211.1 | - | 1 | 1686 | |||
| BM718210.1 | - | 1 | 333 | |||
| EU668356.1 | - | 4 | 2824 | |||

ZNF26
Genomic Map