siRNA Details
| Gene Symbol: SLC2A4RG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020062.3 | NP_064446.2 | 8 | 2246 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC2A4RG| Gene Symbol: SLC2A4RG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020062.3 | NP_064446.2 | 8 | 2246 | ||