siRNA Details
| Gene Symbol: CTNNBIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012329.1 | NP_001012329.1 | 5 | 993 | ||
| NM_020248.2 | NP_064633.1 | 6 | 1027 | |||
| XM_017001853.1 | XP_016857342.1 | 6 | 1252 | |||
| XM_017001854.1 | XP_016857343.1 | 6 | 1126 | |||
| XM_017001855.1 | XP_016857344.1 | 5 | 969 | |||
| GenBank mRNA | AB021262.1 | - | 6 | 993 | ||
| BC014300.1 | - | 5 | 981 | |||
| BC146892.1 | - | 4 | 715 | 9056820 | ||
| BC146896.1 | - | 4 | 715 | 9056824 | ||

CTNNBIP1
Genomic Map