siRNA Details
| Gene Symbol: CTNNBIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012329.1 | NP_001012329.1 | 5 | 2403 | ||
| NM_020248.2 | NP_064633.1 | 6 | 2437 | |||
| XM_017001853.1 | XP_016857342.1 | 6 | 2662 | |||
| XM_017001854.1 | XP_016857343.1 | 6 | 2536 | |||
| XM_017001855.1 | XP_016857344.1 | 5 | 2379 | |||
| GenBank mRNA | AB021262.1 | - | 6 | 2403 | ||

CTNNBIP1
Genomic Map