siRNA Details
| Gene Symbol: SLC17A7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020309.3 | NP_064705.1 | 12 | 2370 | ||
| GenBank mRNA | AB032436.1 | - | 12 | 2219 | ||
| AK091061.1 | - | 8 | 1539 | |||
| AK294226.1 | - | 12 | 2176 | |||
| AK294405.1 | - | 12 | 2219 | |||
| BC059379.1 | - | 12 | 2345 | 5262415 | ||

SLC17A7
Genomic Map