siRNA Details
| Gene Symbol: PBXIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317734.1 | NP_001304663.1 | 9 | 1737 | ||
| NM_001317735.1 | NP_001304664.1 | 7 | 1406 | |||
| NM_020524.3 | NP_065385.2 | 10 | 1826 | |||
| GenBank mRNA | AF221521.1 | - | 10 | 1775 | ||
| AK022497.1 | - | 9 | 1696 | |||
| AK300544.1 | - | 6 | 594 | |||
| AK300987.1 | - | 6 | 1115 | |||
| AK303413.1 | - | 7 | 1350 | |||
| AK303863.1 | - | 7 | 1251 | |||
| AK315551.1 | - | 10 | 1806 | |||
| BC016852.1 | - | 10 | 1754 | 3900461 | ||
| U00952.1 | - | 1 | 295 | |||

PBXIP1
Genomic Map